Would you like to log in again?
Hema-Screen® Lab Pack, Occult Blood Tests, Stanbio Laboratory
Supplier: Stanbio Laboratory
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Hema-Screen®
1290-100
10152-708EA 86.45 USD InStock
10152-708
Hema-Screen® Lab Pack, Occult Blood Tests, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Hema-Screen® Lab Pack is for the qualitative detection of fecal occult blood and uses the guiac test format.
Certifications: FDA cleared, CE marked and Health Canada Licensed.
Ordering information: Each test kit comes complete with Hema-Screen® developing solution, simple-to-follow patient instructions, wooden applicators and a technical insert.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty