Would you like to log in again?
PVAmicro Cloth, Vileda Professional-FHP
Supplier: Vileda Professional
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
143587 143588 143585 143586
10153-212CS 204.96 USD InStock
10153-212 10153-210 10153-220 10153-218
PVAmicro Cloth, Vileda Professional-FHP
Wipes Reusable Cleaning Cloths Cloths for General Applications
PVAmicro is an innovative all-purpose cloth made from microfiber fabric impregnated with PVA.
  • PVAmicro holds 40 times less particle residue after rinsing, compared to ordinary knitted microfibre cloth
  • Cloth can be washed repeatedly
  • Ideal for cleaning all types of hard surfaces, such as office furniture, glass, or stainless steel
The PVA impregnation makes the cloth very easy to rinse and wring. PVAmicro achieves a perfect, shiny cleaning result without streaks every time.
Ordering information: PVAmicro cloth is packaged 5 cloths per pack, 20 packs per case.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty