Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Mini Hotplate Stirrer, 120 V

Supplier: VWR
VWR®
80863881
10153-308EA 590.7 USD InStock
10153-308
VWR® Mini Hotplate Stirrer, 120 V
Stirrers Magnetic Hotplate Stirrers
Rugged, compact hotplate stirrers feature an ergonomic design and are ideal for educational labs.

  • Rugged 5x5" Aluminum Top Plate
  • Boils 300 mL of Water in 16 minutes
  • Built-In Support Rod Holder
  • Five-Year Warranty on Parts and Labor

These units can heat and stir up to 1000 mL of water

The built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Kit includes 45.7 cm (18”) support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector. Optional support plate allows hot plate to be mounted to lab frames or ring stands.

Durable 5x5" cast aluminum top plate will not chip or crack, and provides a uniform heating surface. Bi-metallic thermostat offers reliable temperature control. Powerful heater reaches maximum temperature in minutes. Powerful motor and magnet deliver reliable and consistent stirring.

Certifications: TUV listed. CE marked. RoHS certified.

Ordering information: The 120V units include a 183 cm (72"), 3-wire cord and plug. All units include a five-year limited warranty on parts and labor.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty