Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Low Profile Magnetic Stirrer
Supplier: VWR
VWR®
MS-01FU-VWR
10153-690EA 241.34 USD InStock
10153-690
VWR® Low Profile Magnetic Stirrer
Stirrers Magnetic Stirrers
The VWR® Low Profile Magnetic Stirrer is a laboratory staple.
  • Compact and sleek design
  • Variable Speed: 15 to 1500 rpm
  • Designed with a high level of waterproofing protection
  • VWR® 2 Year Warranty
The small footprint and sleek design preserve valuable bench space, while the unit provides excellent stirring capability for many routine applications.
Ordering information: Magnetic stir bars are sold separately (VWR® SpinBar® 58948-984 is recommended for use with this unit.).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty