Would you like to log in again?
Centrifuge Tube, Polypropylene, Globe Scientific
Supplier: Globe Scientific
6241
10153-796CS 322.3 USD InStock
10153-796
Centrifuge Tube, Polypropylene, Globe Scientific
Tubes Centrifuge Tubes
Centrifuge tubes feature flat-top, plug-seal, leak-resistant screw caps, a large white writing area, and printed graduation marks.
  • RNase/DNase and pyrogen free
  • Cap is made from High Density Polyethylene (HDPE)
  • Spin rate: 10000 RCF (12,320 ×g)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty