Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Vortex Mixers, 230 V and European plugs (Export Only)

Supplier: VWR
VWR®
80862948 9453VWPSEUC 80862952 80862976 80862943 9453VWDGEUC
10153-840EA 410.75 USD InStock
10153-840 10153-818 10153-836 10153-814 10153-822 10153-810
VWR® Vortex Mixers, 230 V and European plugs (Export Only)
Shakers and Mixers Vortex Mixers
Fixed high speed mixing and variable speed models.

  • 230 V model
  • Five year warranty for parts and labor
  • All models include a cup head and three inch platform
  • Manufactured in the USA

VWR® Fixed Speed Vortex Mixers start mixing when the cup head is depressed. Speed is a full 3200 rpm to provide vigorous mixing of samples.

VWR® Analog Vortex Mixers offer variable analog speed control with continuous or touch mode operation. Control allows low rpm start up for gentle shaking or high speed mixing for vigorous vortexing of samples. Two modes of operation; continuous mode when using optional accessory attachments or touch mode which activates mixing when depressing the cup head.

VWR® Digital Vortex Mixers are ideal for applications that demand repeatable results. Microprocessor control maintains set speed for strong, consistent mixing action. Touch pad controls and independent LED displays for speed and time. Timer will display elapsed time or when programmed to a user defined time limit, the unit will shut off when time reaches zero.Two modes of operation; continuous mode when using the optional accessory attachments, or touch mode which activates mixing when depressing the cup head.

VWR® Pulsing Vortex Mixers are ideal for glass bead cell disruption/homogenization and provide a powerful pulsing vortex action.Capable of complete cell disruption of samples in only minutes.Unique pulsing action mixes at maximum speed for 30 seconds and rests for ten seconds reducing heat generation while providing more effective mixing and disruption. Displayed time counts up during continuous operation and counts down during timed runs. Also includes a stainless steel microtube holder with built-in cup head.

Certifications: TUV listed, CE Marked. RoHS compliant.

Ordering information: All models include a cup head, three-inch head with cover, and a 72" (183 cm) cord with a Euro style plug. Pulsing Vortex Mixer includes a stainless steel microtube holder with built-in cup head and holds up to (12) 1.5 to 2 ml microtubes. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.

Caution: 230 V model.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty