Would you like to log in again?
Multiparameter Water Quality Meters, smarTROLL, In-Situ
Supplier: IN-SITU, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
smarTROLL
71970 72000 71980 71990
10153-862EA 6169.77 USD InStock
10153-862 10153-860 10153-866 10153-864
Multiparameter Water Quality Meters, smarTROLL, In-Situ
Electrochemical Meters pH and Multi-Parameter Meters
The smarTROLL Multiparameter Handheld combines optical dissolved oxygen (DO) technology with revolutionary smartphone mobility
Simultaneously measure 14 parameters: Dissolved oxygen with the optical RDO Sensor, Conductivity (actual and specific), pH, ORP, Salinity, Total dissolved solids, Resistivity, Density, Barometric pressure, Air temperature and Water temperature. Get instant results—No warm-up or stabilization time required.
Waterproof case is availabe for your iPhone and iPod touch device.
The intuitive iSitu® application eliminates training time and runs on your iPhone®, iPod touch®, or iPad® device.
Certifications: FCC and EPA Approved. CE compliant.
Ordering information: Bundles include multiparameter probe, weighted guard, battery pack, specified cable length, calibration cup and Operator’s Manuals on CD. Does not include iOS device.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty