Would you like to log in again?
Hygiena SpotCheck Plus™, Hygiene Non-Instrument Color Tests, Hardy Diagnostics
Supplier: Hardy Diagnostics
SpotCheck Plus™
GL100 GL100
10154-026BX 362.91 USD InStock
CA10154-026 10154-026
Hygiena SpotCheck Plus™, Hygiene Non-Instrument Color Tests, Hardy Diagnostics
Monitoring Systems Hygiene Monitoring Systems
A rapid non-instrument hygiene monitoring test utilizing the snap-valve™ technology.
  • Detects residual lactose and glucose on surfaces
  • Results in 60 minutes
A rapid non-instrument hygiene monitoring test utilizing the snap-valve™ technology. The reagent in the swab detects residual lactose and glucose on surfaces resulting in a color change. Results in 60 seconds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty