Would you like to log in again?
All-Purpose Cable Lockout, with 8' Sheathed Cable, Brady Worldwide®
Supplier: Brady
50943 50943
10154-258EA 69.93 USD InStock
10154-258 CA89079-570
All-Purpose Cable Lockout, with 8' Sheathed Cable, Brady Worldwide®
Security Locks
Cable lockouts are incredibly versatile, making them perfect for any unusual energy isolating devices that are difficult to lock out
Compact, lightweight body is made of rugged, impact modified glass filled nylon giving them superior chemical, corrosion, and temperature resistant properties. Sheathed metal cable ha as 3/16" diameter, including vinyl coating. Ergonomic design makes it a "cinch" to tighten the cable; simply hold device in one hand and cinch tight with the other. Built-in cable loop keeps cable neatly coiled when stored and prevents it from dangling loose when device is in use. The four lock holes in handle accommodate most lock shackles and hasps. Additional 8', 12', 16', and 20' cables are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty