Would you like to log in again?
Thermo Scientific™ MAS™ Diabetes Controls, Thermo Scientific
Supplier: Thermo Scientific
MAS™
DBCL-V DBCL-MP
10154-276PK 831.06 USD InStock
10154-276 10158-928
Thermo Scientific™ MAS™ Diabetes Controls, Thermo Scientific
Control reagents
Monitor hemoglobin A1c methods with MAS™ Diabetes Control, a bi-level control available in a convenient Vista™ vial format
Human whole blood matrix. Assayed values available for most major analyzers.
Analytes: Hemoglobin A1c.
This read-to-use, liquid assayed diabetes control has been assayed for most major instrument systems.
Ordering information: Thermo Scientific™ MAS™ Diabetes Controls, Thermo Fisher Scientific
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty