Would you like to log in again?
Capto™ S ImpAct Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Capto™
17-3717-02 17-3717-01 17-3717-01 17-3717-02
10158-348EA 815.54 USD InStock
CA10158-346 10158-348 10158-346 CA10158-348
Capto™ S ImpAct Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Capto™ S ImpAct is a strong cation exchange chromatography medium (resin) developed for polishing of monoclonal antibodies (MAbs) and other biomolecules.
  • High binding capacity, >100 mg MAb/ml medium
  • Efficient aggregate removal at high load of monoclonal antibodies
  • High resolution polishing based on the well-established Capto™ platform
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimisation and scaling
  • High productivity enabling cost-efficient manufacturing
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty