Would you like to log in again?
HiTrap Columns, Capto S ImpAct, Cytiva
Supplier: Cytiva
HiTrap™
17-3717-55 17-3717-55 17-3717-51 17-3717-51
10158-352EA 1264.39 USD InStock
CA10158-352 10158-352 CA10158-350 10158-350
HiTrap Columns, Capto S ImpAct, Cytiva
Chromatography Columns
HiTrap Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium for screening and small-scale protein purifications using ion exchange chromatography (IEX).
  • Convenient prepacked column for fast, reproducible, and easy small-scale purifications
  • High binding capacity, >100 mg MAb/ml medium
  • Efficient aggregate removal at high load of monoclonal antibodies
  • High-resolution intermediate purification and polishing based on the well-established Capto™ platform
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty