Would you like to log in again?
GelJacket™ Stackable CO₂ Incubators, 7404 Series, Caron Products
Supplier: Caron Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7404-10-3 7404-10-1 7404-10-2
10158-852EA 25015.65 USD InStock
10158-852 10158-772 10158-770
GelJacket™ Stackable CO₂ Incubators, 7404 Series, Caron Products
Incubators CO₂ Incubators
Caron’s GelJacket is a high-volume, controlled humidity CO2 incubator that replaces conventional water jacketed units as the benchmark for critical culture protection.
GelJacket offers precise gVapor™ controlled humidification, as well as a rapid 90° decon cycle for fast chamber cleaning. These standard features save energy and user time, as opposed to contamination-prone water pan-based systems and high temperature or complicated competing decontamination processes.
Refrigerated models with extended temperature ranges are available for applications that generate heat (such as shakers). A wide range of accessories are available to allow you to customize the incubator for your specific application.
Note: View the Product Accessories List, in Datasheets under Related Information. To order specific accessories, search by part number.
Caron's proprietary thermal gel insulation provides power outage temperature protection equivalent to water, with minimal maintenance, and at a fraction of the weight. Leaks and regular jacket fillings are now a thing of the past.
Ordering information: Accessories available separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product.
Extended Warranty Packages offer peace of mind with up to an additional 1 year or 3 years on top of the original warranty, covering parts and labor for any manufacturer's issues that may occur during this time. Purchase of an Extended Warranty is only valid prior to the original warranty expiration and is only available for U.S. Customers. See table below for more information.
PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.
PM Service Kits for Accessories help to minimize downtime by providing routine maintenance items for accessories. See table below for more information.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty