Would you like to log in again?
Dehydrant Blends, Epredia
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Richard-Allan Scientific®
6401 6355 6415 6355 6515 6401 6201 6215 6501 6301 6301 6315 6201 6501
10128-124CS 199.15 USD InStock
10128-124 10128-122 10158-872 10158-870 84000-050 66004-948 CA66004-946 CA66004-948 66004-946 CA84000-050 CA84000-046 84000-048 84000-046 CA84000-048
Dehydrant Blends, Epredia
Stains Microscopy and staining reagents
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty