Would you like to log in again?
Screw Top Pathology Containers, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ES60240 ES60180 ES60480 ES61000 ES60120
10159-048CS 84.35 USD InStock
10159-048 10159-044 10159-050 10159-040 10159-042
Screw Top Pathology Containers, Azer Scientific
Sample Containers General Purpose Containers
Manufactured from high-quality, clarified polypropylene and resistant to histology reagents.
  • High-Quality, High-Density Polyethylene
  • Screw Top Lid
  • Leak-Resistant Closure
  • Histology Reagent Resistant
The screw top lid provides a leak-resistant closure to help protect the specimen and increase safety for lab personnel.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty