Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

XBF40D-MD –40 °C Blast Freezer for Plasma Freezing, Thermo Scientific

Supplier: Thermo Scientific
XBF40D-MD
10159-058EA 136999.97 USD InStock
10159-058
XBF40D-MD –40 °C Blast Freezer for Plasma Freezing, Thermo Scientific
Freezers Laboratory / General Purpose Freezers
The Thermo Scientific XBF40D-MD Blast Freezer is deisgned for plasma freezing applications

The freezer provides a total capacity of 815.5L (28.8cu.ft.), with a 419L (14.8cu.ft.) freezing area. Freezer temperature is preset at –40°C. Cabinet material for these upright units is cold-rolled steel. Freezers are equipped with auto defrost.

Certifications: C-UL listed.

Delivery information: Unit is supplied with three stainless steel shelves, six aluminum carrying baskets, and one wireless triple timer.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty