Would you like to log in again?
VWR® Tube Rocker Collection
Supplier: VWR
280150V 260450V 282000V
10159-752EA 729.33 USD InStock
10159-752 10159-754 10159-756
VWR® Tube Rocker Collection
Shakers and Mixers Rotating/Rocking Shakers
The VWR® Tube Rocker Collection features Tube Rockers available in a "Mini" size as well as an "XL" size and in Fixed Speed or a Variable Speed models.
  • Low profile and small footprint
  • Tube Rockers provide smooth, gentle mixing
  • Available with a variety of speed settings
The Mini Tube Rockers holds 8 tubes, 120 mm long while the XL model holds up to 18 tubes, also 120 mm long. Each model is equipped with a fixed tilt angle of 35 degrees and mixes your samples simultaneously via a 110-240V power pack. The variable speed models are adjustable in 1 rpm increments from 5-25 rpm while the fixed speed model is fixed at 18 rpm's.
Certifications: 12V DC via AC Adapter carries UL/CSA and CE approvals.
Ordering information: Each unit comes with a 2-year warranty and is supplied with a 12V AC Adapter for use both internationally and domestically.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty