Would you like to log in again?
Plastic Littauer Suture Scissors, Sklar
Supplier: Sklar
96-2905 96-2906
10184-674EA 287.68 USD InStock
10184-674 10160-130
Plastic Littauer Suture Scissors, Sklar
Scissors
Littauer Suture Scissors are a basic instrument used for suture removal
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer suture scissors typically have larger, less delicate tips than Spencer Stitch Scissors, and are often used for removing larger sutures. They should not be used for cutting tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty