Would you like to log in again?
Plastic Dressing Forceps, Disposable Grade, Sklar
Supplier: Sklar
96-2916
10160-136BX 66.98 USD InStock
10160-136
Plastic Dressing Forceps, Disposable Grade, Sklar
Forceps
Plastic Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds during laser surgeries
This product is straight with serrated tips and a length of 5 inches. It is non-sterile and sold in boxes of 100.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty