Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wrist Action Shakers, 220 V

Supplier: Burrell Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Wrist-Action®
075-775-16-36 075-775-08-36 075-765-16-36 075-775-04-36
10161-188EA 9053.03 USD InStock
10161-188 10161-178 10161-180 10161-184
Wrist Action Shakers, 220 V
Shakers and Mixers Rotating/Rocking Shakers
Wrist action shaker, with 385 oscillations per minute (+/-) 10%.

  • 220 V model

Max. load tested at 10 lbs. blue/white color.

Certifications: Complies with UL, CSA and CE.

Ordering information: Each wrist shaker comes with enough clamps to accommodate its full capacity of vessels. Optional top platform accessory accommodates up to eight 250 ml flasks and attaches directly to main shaft of shaker.

Caution: 220 V model. Corresponding models with voltage suitable for use in US (10161-186, 57040-027, 57040-049, 57040-060, 57040-082) are also available, please see separate product page.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty