Would you like to log in again?
Plastic Procedure Trays, Perforated, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10-1966-6 10-1966 10-1965 10-1965-6 10-1967 10-1965-12 10-1966-12 10-1967-6 10-1967-12
10193-480PK 367.05 USD InStock
10193-480 10193-490 10193-484 10193-482 10193-488 10193-486 10163-412 10163-482 10193-478
Plastic Procedure Trays, Perforated, Sklar®
Trays/Dishes/Pots Instrument Trays/Dishes
Plastic Procedure Tray Perforated
Reusable Plastic. Autoclavable. Features No-Slip Textured Surface.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty