Would you like to log in again?
Posi-Locking Instrument Holders, OR Grade, Sklar
Supplier: Sklar
10-1337 10-1334 10-1330 10-1336 10-1332 10-1338 10-1339
10163-524EA 454.1 USD InStock
10163-524 10163-526 10163-520 10163-522 76191-682 10163-906 10163-908
Posi-Locking Instrument Holders, OR Grade, Sklar
Forceps and Instrument Holders
These Posi-Locking Instrument Holders are used to hold instruments such as hemostatic forceps, needle holders, and scissors in the open position during sterilization.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Once aligned, a locking mechanism ensures the instruments do not slide off the holder. While the locking mechanism provides secure placement of the instruments during cleaning, transporting, and storage, it makes removal of the instruments more difficult when preparing for a procedure than if a non-locking instrument stringer were used. Multiple sizes of the Posi Locking holder are available depending on the volume of instruments being used.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty