Would you like to log in again?
Instrument Stringers, OR Grade, Sklar
Supplier: Sklar
10-1348 10-1346 10-1342 10-1340 10-1344 10-1347
10163-528EA 191.89 USD InStock
10163-528 10163-508 10163-534 10163-530 10163-862 10163-532
Instrument Stringers, OR Grade, Sklar
Forceps and Instrument Holders
Instrument stringers are used to hold instruments during cleaning and transport them to storage and keep them organized.
  • Made of stainless steel
Unlike the Posi lock instrument holder, there is no locking mechanism on this instrument stringer. This allows for ease of removing the instruments, but also may result in contaminated instruments should they slide off the stringer prior to use in a procedure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty