Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SklarLite™ Full Size Rigid Sterilization Container System, Sklar
Supplier: Sklar
SklarLite™
10-4112 10-4030 10-4146 10-4110 10-4048 10-4058 10-4170 10-4042 10-4066 10-4088 10-4016 10-4080 10-4070 10-4092 10-4004 10-4236 10-4002 10-4210 10-4200 10-4138 10-4176 10-4082 10-4000 10-4164 10-4222 10-4010 10-4232 10-4160 10-4150 10-4132 10-4072 10-4012 10-4226 10-4192 10-4094 10-4038 10-4086 10-4238 10-4178 10-4064 10-4126 10-4198 10-4068 10-4202 10-4152 10-4114 10-4006 10-4100 10-4182 10-4208 10-4168 10-4154 10-4148 10-4184 10-4144 10-4162 10-4108 10-4196 10-4034 10-4172 10-4156 10-4130 10-4022 10-4190 10-4040 10-4090 10-4056 10-4014 10-4122 10-4134 10-4028 10-4078 10-4026 10-4180 10-4052 10-4224 10-4128 10-4054 10-4102 10-4062 10-4220 10-4166 10-4096 10-4116 10-4118 10-4204 10-4001 10-4036 10-4018 10-4136 10-4046 10-4084 10-4214 10-4194 10-4188 10-4020 10-4228 10-4142 10-4050 10-4032 10-4234 10-4074 10-4076 10-4158 10-4216 10-4060 10-4024 10-4174 10-4140 10-4044 10-4104 10-4098 10-4124 10-4120 10-4218 10-4008 10-4230 10-4206 10-4212
10163-590EA 1418.01 USD InStock
10163-590 10163-912 10163-638 10163-914 10163-634 10163-910 10163-636 10163-596 10163-630 10163-598 10163-632 10163-592 10163-594 10163-580 10163-626 10163-628 10163-622 10163-986 10163-624 10163-988 10163-586 10163-982 10163-620 10163-984 10163-582 10163-980 10163-584 10163-690 10163-608 10163-978 10163-618 10163-578 10163-732 10163-974 10163-612 10163-734 10163-976 10163-614 10163-970 10163-574 10163-696 10163-576 10163-972 10163-730 10163-698 10163-610 10163-692 10163-572 10163-694 10163-718 10163-604 10163-968 10163-726 10163-606 10163-728 10163-688 10163-600 10163-722 10163-964 10163-966 10163-724 10163-684 10163-960 10163-686 10163-962 10163-720 10163-680 10163-682 10163-708 10163-956 10163-714 10163-958 10163-716 10163-952 10163-710 10163-678 10163-712 10163-954 10163-674 10163-950 10163-676 10163-670 10163-672 10163-938 10163-704 10163-946 10163-948 10163-706 10163-666 10163-942 10163-700 10163-668 10163-944 10163-702 10163-662 10163-664 10163-940 10163-660 10163-928 10163-934 10163-936 10163-930 10163-656 10163-932 10163-658 10163-652 10163-654 10163-650 10163-916 10163-918 10163-648 10163-924 10163-926 10163-644 10163-920 10163-646 10163-922 10163-640 10163-642
SklarLite™ Full Size Rigid Sterilization Container System, Sklar
Boxes Transport Boxes
The SklarLite™ Rigid Sterilization Container System provides an organized, routine, and secure approach to the sterilization of surgical instruments
Customize this SklarLite™ System by selecting colored lids and ID labels. Available in a choice of six vibrant colors for easy indentification of instrument sets by procedure and/or department. Choose from various configurations to fit surgery center needs.
Accessories information: SklarLite™ Identification Labels are ideally suited for use with the SklarLite™ Rigid Sterilization Containers and Container Systems. Polyester film labels are nonsterile and latex-free. Disposable identification labels are color coded to enable quick and simple sorting and selection.
The SklarLite™ endoscopy rack is part of an overall sterilization system to be used with sterilization containers. Sklar® also offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty