Would you like to log in again?
SklarLite™ Full Size Rigid Sterilization Container Set, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SklarLite™
10-4248 10-4280 10-4258 10-4260 10-4240 10-4254 10-4272 10-4256 10-4276 10-4266 10-4246 10-4270 10-4244 10-4274 10-4268 10-4252 10-4278 10-4037 10-4262 10-4250 10-4282 10-4242 10-4025 10-4264 10-4013
10163-736EA 577.27 USD InStock
10163-736 10163-758 10163-616 10163-738 10163-776 10163-754 10163-756 10163-778 10163-750 10163-772 10163-752 10163-774 10163-770 10163-748 10163-744 10163-766 10163-602 10163-768 10163-746 10163-762 10163-740 10163-742 10163-764 10163-588 10163-760
SklarLite™ Full Size Rigid Sterilization Container Set, Sklar
Boxes Transport Boxes
The SklarLite™ Rigid Sterilization Container Set provides an organized, routine, and secure approach to the sterilization of surgical instruments.
  • Sterilization container lid is available in a variety of colours
  • Lids are made of anodized aluminium alloy, these reusable lids are nonsterile and latex-free
Similar to the SklarLite™ Full Size Rigid Sterilization Container Systems, these containers are customizable with the use of colored lids and ID labels. These Full Size Container Sets include additional organizational tools, including a 10.2 cm (4") wire basket.
Accessories information: SklarLite™ Identification Labels are ideally suited for use with the SklarLite™ Rigid Sterilization Containers and Container Systems. Polyester film labels are nonsterile and latex-free. Disposable identification labels are color coded to enable quick and simple sorting and selection.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty