The SklarLite™ Rigid Sterilization Container provides an organized, routine, and secure approach to the sterilization of surgical instruments
Customize the SklarLite™ container by selecting colored lids and ID labels. Available in a choice of six vibrant colors for easy indentification of instrument sets by procedure and/or department. Choose from various configurations to fit surgery center needs.
Accessories information: SklarLite™ Identification Labels are ideally suited for use with the SklarLite™ Rigid Sterilization Containers and Container Systems. Polyester film labels are nonsterile and latex-free. Disposable identification labels are color coded to enable quick and simple sorting and selection.