Would you like to log in again?
Omega Supreme Cleanroom HEPA Vac, Atrix
Supplier: Atrix
VACOMEGASECRH
101640-742EA 656.92 USD InStock
101640-742
Omega Supreme Cleanroom HEPA Vac, Atrix
Vacuum Cleaners
The Omega Supreme Cleanroom 1 Gallon HEPA dry particulate vacuum offers HEPA filtered exhaust air to reduce carbon particle disbursion from the motor brushes
Replacement Filters are designed to capture fumes, atmospheric dust, black toner and all color toner and other ultra-fine particulate. Compatible with 3M vacuums.
And, at less than 70 decibels, it gets the job done quietly, quickly and cleanly with EMI/RFI Line Noise Filter. HEPA Efficiency: 99.97% @ 0.3 microns. Exceeds Class 100,000.
Ordering information: Includes detachable power cord, 10' ESD safe hose, ESD goose neck, utility nozzle, shoulder strap
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty