Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BL21 (DE3) Competent E. coli, New England Biolabs
Supplier: New England Biolabs
C2527I C2527I C2527H C2527H
101640-792EA 177 USD InStock
101640-792 101640-790 CA101640-792 CA101640-790
BL21 (DE3) Competent E. coli, New England Biolabs
Competent Cells
Chemically competent E.coli cells suitable for transformation and protein expression.
A widely used T7 expression E. coli strain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty