Would you like to log in again?
PUREXPRESS® delta Ribosome Kit, New England Biolabs
Supplier: New England Biolabs
E3313S E3313S
101640-812EA 393 USD InStock
CA101640-812 101640-812
PUREXPRESS® delta Ribosome Kit, New England Biolabs
Protein Expression Protein Expression Systems
A rapid method for gene expression analysis, PURExpress® is a novel cell-free transcription/translation system reconstituted from the purified components necessary for E
The relative nuclease-free and protease-free nature of the PURExpress platform preserves the integrity of DNA and RNA templates/ complexes and results in proteins that are free of modification and degradation. Transcription and translation are carried out in a one-step reaction, and require the mixing of only two tubes. With results available in a few hours, PURExpress saves valuable laboratory time and is ideal for high throughput technologies. The ribosomes are removed from this kit, allowing users to add their own modified ribosomes for studies on protein translation. Control ribosomes (sufficient for 2 reactions) are provided in a separate tube.
coli translation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty