Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBNEXT® End Repair Module, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E6050L E6050L E6050S E6050S
101640-832EA 414 USD InStock
CA101640-834 101640-832 CA101640-832 101640-834
NEBNEXT® End Repair Module, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext® End Repair Module has been optimized to repair DNA fragmented by nebulization, acoustic shearing, or nucleases
Fragmented DNA is converted by the NEBNext End Repair Module to blunt ended DNA having 5´-phosphates and 3´-hydroxyl. The NEBNext End Repair Module is provided as a master mix to maximize efficiency and convenience in DNA sample preparation workflows.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty