Would you like to log in again?
PUREXPRESS® Disulfide Bond Enhancer, New England Biolabs
Supplier: New England Biolabs
E6820S E6820S
101640-914EA 279 USD InStock
101640-914 CA101640-914
PUREXPRESS® Disulfide Bond Enhancer, New England Biolabs
Protein Expression Protein Expression Systems
The PURExpress® Disulfide Bond Enhancer (PDBE) is a proprietary blend of proteins and buffer components designed to correctly fold target proteins with multiple disulfide bonds produced in PURExpress reactions or E.coli S30 extracts.
Added at the beginning of a reaction, the components promote a proper disulfide bond pattern by assisting with the oxidation of cysteine thiols and correcting mis-oxidized substrates. These enhancements can increase the yield of soluble and functionally active protein.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty