Would you like to log in again?
hSMUG1
Supplier: New England Biolabs
M0336S M0336S
101640-950EA 85 USD InStock
101640-950 CA101640-950
hSMUG1
Enzymes
Human single-strand-selective monofunctional uracil-DNA Glycosylase (hSMUG1) excises deoxyuracil and deoxyuracil-derivatives bearing an oxidized group at C5, such as 5-hydroxyuracil, 5-hydroxymethyluracil and 5-formyluracil in ssDNA and dsDNA.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty