Would you like to log in again?
T4 Phage Beta-Glucosyltransferase, New England Biolabs
Supplier: New England Biolabs
M0357L M0357L M0357S M0357S
101640-964EA 410 USD InStock
CA101640-966 101640-964 CA101640-964 101640-966
T4 Phage Beta-Glucosyltransferase, New England Biolabs
Enzymes
T4 Phage β-glucosyltransferase specifically transfers the glucose moiety of uridine diphosphoglucose (UDP-Glc) to the 5-hydroxymethylcytosine (5-hmC) residues in double-stranded DNA, making beta-glucosyl-5-hydroxymethylcytosine.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty