Would you like to log in again?
ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Supplier: New England Biolabs
ONETAQ® QUICK-LOAD®
M0486S M0486L M0486S M0486L
101640-998EA 50 USD InStock
CA101640-996 101640-998 101640-996 CA101640-998
ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq® Quick-Load® 2X Master Mix with Standard Buffer is an optimized, ready-to-use blend of Taq and Deep VentR ™ DNA Polymerases ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products.
The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient quick-load master mix formulation contains dNTPs, MgCl2, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask comigrating DNA bands.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty