Would you like to log in again?
ONETAQ® Hot Start QUICK-LOAD® 2X Master Mix with GC Buffer, New England Biolabs
Supplier: New England Biolabs
ONETAQ® QUICK-LOAD®
M0489L M0489S M0489S M0489L
101641-010EA 83 USD InStock
CA101641-010 101641-010 CA101641-008 101641-008
ONETAQ® Hot Start QUICK-LOAD® 2X Master Mix with GC Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq® Hot Start Quick-Load® 2X Master Mix with GC Buffer is an optimized, ready-to-use blend of Taq and Deep VentR™ DNA Polymerases combined with an aptamer-based inhibitor.
This enzyme blend is ideally suited to PCR applications from GC-rich templates, including pure DNA solutions, bacterial colonies, and cDNA products. The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgSO4, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask co-migrating DNA bands.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty