Would you like to log in again?
Phusion High-Fidelity PCR Master Mix with HF® Buffer, New England Biolabs
Supplier: New England Biolabs
Phusion®
M0531L M0531S M0531S M0531L
101641-016EA 832 USD InStock
CA101641-016 101641-016 CA101641-018 101641-018
Phusion High-Fidelity PCR Master Mix with HF® Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion High-Fidelity PCR Master Mix with HF Buffer is a 2X master mix consisting of Phusion DNA Polymerase, deoxynucleotides and reaction buffer that has been optimized and includes MgCl2
All that is required is the addition of template, primers and water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty