Would you like to log in again?
dATP Solution, New England Biolabs
Supplier: New England Biolabs
N0440S N0440S
101641-036EA 63 USD InStock
CA101641-036 101641-036
dATP Solution, New England Biolabs
Nucleic Acid Reagents Nucleotides
dAtp Solution contains 0.25 ml of a 100 mM solution of ultrapure dATP as a sodium salt at pH 7.4.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty