Would you like to log in again?
Ribonucleotide Solution Set, New England Biolabs
Supplier: New England Biolabs
N0450L N0450S N0450L N0450S
101641-040EA 82 USD InStock
101641-040 CA101641-038 101641-038 CA101641-040
Ribonucleotide Solution Set, New England Biolabs
Nucleic Acid Reagents Nucleotides
The Ribonucleotide Solution Set (NTP Set) consists of four separate solutions of ATP, GTP, CTP and UTP, pH 7.5, as sodium salts
Each nucleotide is supplied at 100 mM. The NTPs are also available as a Ribonucleotide Solution Mix.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty