Would you like to log in again?
KpnI-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R3142M R3142S R3142S R3142L R3142M R3142L
101641-150EA 309 USD InStock
CA101641-152 101641-150 101641-152 CA101641-148 101641-148 CA101641-150
KpnI-HF®, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that carries the cloned and modified KpnI gene from Klebsiella pneumoniae OK8 (ATCC 49790).
High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty