Would you like to log in again?
SacI-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R3156M R3156M
101641-164EA 287 USD InStock
CA101641-164 101641-164
SacI-HF®, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that carries the cloned and modified SacI gene from Streptomyces achromogenes (ATCC 12767).
High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty