Would you like to log in again?
Casein Kinase II
Supplier: New England Biolabs
P6010S P6010L P6010S P6010L
101641-526EA 714 USD InStock
CA101641-526 101641-526 CA101641-528 101641-528
Casein Kinase II
Enzymes
Casein Kinase II (CK2) is a constitutively active serine/threonine protein kinase composed of two 44 kDa catalytic α-subunits and two 26 kDa regulatory β-subunits in an α2β2 configuration to form stable heterotetramers
CK2 holoenzyme undergoes autophosphorylation at two serine residues (S2/S3) of its β-subunit. Recently it has been shown that CK2 α-subunits undergo intermolecular tyrosine-autophosphorylation at Y182, which may represent a specific regulatory mechanism. Also, CK2 is able to phosphorylate, under special circumstances, tyrosyl residues in proteins. CK2 is implicated in a variety of cellular functions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty