Would you like to log in again?
RNA Loading Dye (2X), New England Biolabs
Supplier: New England Biolabs
B0363S B0363S
101641-556EA 59 USD InStock
CA101641-556 101641-556
RNA Loading Dye (2X), New England Biolabs
Stains Electrophoresis Stains
The RNA Loading Dye, (2X) is a premixed loading dye for use with denaturing and non-denaturing PAGE/agarose gels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty