Would you like to log in again?
PUREXPRESS® (aa tRNA) Kit, New England Biolabs
Supplier: New England Biolabs
E6840S E6840S
101641-562EA 408 USD InStock
CA101641-562 101641-562
PUREXPRESS® (aa tRNA) Kit, New England Biolabs
Protein Expression Protein Expression Systems
A rapid method for gene expression analysis, PURExpress® is a novel cell-free transcription/translation system reconstituted from the purified components necessary for E
The relative nuclease-free and protease-free nature of the PURExpress platform preserves the integrity of DNA and RNA templates/ complexes and results in proteins that are free of modification and degradation. Transcription and translation are carried out in a one-step reaction, and require the mixing of only two tubes. With results available in a few hours, PURExpress saves valuable laboratory time and is ideal for high throughput technologies. The tRNA and amino acids are supplied separately in this kit, allowing users to run a protein synthesis reaction by adding modified amino acids and tRNA mixtures to the reaction.
coli translation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty