Would you like to log in again?
E. coli Pyrophosphatase
Supplier: New England Biolabs
M0361L M0361L M0361S M0361S
101641-570EA 284 USD InStock
CA101641-570 CA101641-572 101641-570 101641-572
E. coli Pyrophosphatase
Enzymes
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate
P207–4 + H20 → 2HP04–2. Pyrophosphatase, Inorganic (E. coli) is prepared from a clone of the E. coli inorganic pyrophosphatase gene.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty