Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
E.coli Ribosome, New England Biolabs
Supplier: New England Biolabs
P0763S P0763S
101641-600EA 211 USD InStock
CA101641-600 101641-600
E.coli Ribosome, New England Biolabs
Competent Cells
The ribosome is the machinery for protein synthesis in a cell
The 70S E. coli Ribosome consists of a small subunit (30S) and a large subunit (50S). This preparation of ribosomes is highly active in NEB’s PURExpress Protein Synthesis Kit, and can be used in ribosome structure and function studies, as a target for drug screening and as starting material for isolation of native ribosomal RNAs (5S, 16S, 23S). E. coli Ribosome is supplied as a 33.3 mg/ml solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty