Would you like to log in again?
Tall Form High Precision Specific Gravity Hydrometers, Kimble Chase
Supplier: DWK Life Sciences
52111-0708 52111-1213 52111-0809 52111-1314 52111-1011 52111-0810 52111-1111 52111-1212 52111-0910 52111-1112 52111-1415 52111-1414
10169-834EA 46.53 USD InStock
10169-834 10169-812 10169-800 10169-836 10169-802 10169-838 10169-804 10169-806 10169-808 10169-832 10169-810 10169-798
Tall Form High Precision Specific Gravity Hydrometers, Kimble Chase
Hydrometers
Serial number is printed on the scale and a certificate of traceability is included
The hydrometer meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy. The certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards. The ballast is made from steel pellets and a binder, and it is free of heavier metals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty