Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adapters, Sanitary to 80 mm Flange, Chemglass

Supplier: Chemglass
CG-1003-B-10
10170-316EA 527.98 USD InStock
10170-316
Adapters, Sanitary to 80 mm Flange, Chemglass
Fittings and Connectors
80 mm adapter to sanitary fitting adapter.

  • Approximate overall length is 2.5"
  • PTFE
  • Manufactured by Chemglass

This adapter provides a convenient way to connect powder charging bags/bulk addition bags and other sanitary fitting accessories to the addition port on Chemglass reaction systems.

Ordering information: Clamp and seals must be purchased separately.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty