Would you like to log in again?
Adapter, Addition Pipe for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1380-31 CLS-1380-51 CLS-1380-40
75780-224EA 385.09 USD InStock
75780-224 75780-226 10170-318
Adapter, Addition Pipe for Bioreactors, Chemglass
Bioreactors
This adapter can be used to add fluids or gases to the bio reactor
The portion of the adapter that is within the reactor has a short drip tip tube that can accept tubing to reach further into the listed capacity reactor. These Adapters are replacement parts for Applikon® Biotechnology and Broadley-James® brands of bioreactors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty