Would you like to log in again?
Adjustable Sampling Tubes for Bioreactors, M10
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CLS-1385-13 CLS-1383-08 CLS-1380-21
75780-150EA 553.98 USD InStock
75780-150 10170-322 10170-320
Adjustable Sampling Tubes for Bioreactors, M10
Tubes Test Tubes
These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level.
Tubing is ¹/₄" OD. Stainless steel components manufactured from 316 L with an electropolished surface finish of 20Ra or better.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty