Would you like to log in again?
Buchner Funnels, Perforated Plates, Chemglass
Supplier: Chemglass
CG-1402-P-03 CG-1402-P-01 CG-1402-P-02
10170-348EA 119.73 USD InStock
10170-348 10170-346 10170-344
Buchner Funnels, Perforated Plates, Chemglass
Funnels Filtration Funnels Büchner Funnels
Buchner funnels having a perforated plate rather than a fritted disc.
Funnels accept filter paper size as listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty